Learn More
Sigma Aldrich Lacey Carbon Supported Copper Grids 200 Mesh 50nm Hole Size 5/box
Catalog No. 503125906 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Pk.
Catalog No. Quantity
50-312-5906 1 Pk.
1 options
Catalog No. 50-312-5906 Supplier Sigma Aldrich Supplier No. TEMLC200CU
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Lacey Carbon Supported Copper Grids 200 Mesh 50nm Hole Size 5/box
Specifications
Quantity 1 Pk.