Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LACRT Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

$209.50 - $511.50

Specifications

Antigen LACRT
Concentration 0.5 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5121805
View Documents
Invitrogen™
PA5121805
20 μL
Each for $209.50
Only null left
Add to Cart
 
PIPA5121804
GREENER_CHOICE
View Documents
Invitrogen™
PA5121804
100 μL
Each for $511.50
Only null left
Add to Cart
 
Description

Description

LACRT Polyclonal antibody specifically detects LACRT in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

LACRT
Western Blot
Unconjugated
Rabbit
Human, Mouse
Q9GZZ8
90070
Glu20-Ala138 (Accession Q9GZZ8), with N-terminal His and GST Tag.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
LACRT
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Extracellular glycoprotein lacritin; lacritin; LACRT
LACRT
IgG
Antigen affinity chromatography, Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.