Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lactacystin
GREENER_CHOICESDP

Catalog No. 2267200U Shop All Novus Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-672-00U 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-672-00U Supplier R&D Systems™ Supplier No. 2267/200U

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cell-permeable, potent and selective proteasome inhibitor

A cell-permeable, potent and selective proteasome inhibitor. A Streptomyces metabolite that is thought to bind irreversibly to the active site N-terminal threonine residue of the catalytic β-subunit of the 20S proteasome, thereby inhibiting its chymotrypsin and trypsin-like activities. Induces neurite outgrowth in Neuro 2a neuroblastoma cells and has been reported to induce apoptosis in human monoblast U937 cells. Also inhibits NF-κB activation (IC50 = 10 μM).

Chemical Identifiers

CAS 133343-34-7
Molecular Formula C15H24N2O7S

Specifications

Quantity 200 μg
Product Line Tocris
Formula Weight 376.42
Purity >98%
Molecular Weight (g/mol) 376.424
Chemical Name or Material Lactacystin
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.