Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lactamase, beta 2 Antibody (OTI2F9), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271901APCCY7

Catalog No. N271901APC7


Only null left
Add to Cart

Description

Description

lactamase, beta 2 Monoclonal specifically detects lactamase, beta 2 in Human, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

lactamase, beta 2
Monoclonal
APC-Cyanine7
PBS with 0.05% Sodium Azide
LACTB2
Recombinant protein fragment corresponding to amino acids 78-288 of human lactamase, beta 2 (NP_057111) produced in HEK293T cells.
0.1 mL
51110.0
Store at 4°C in the dark. Do not freeze.
Flow Cytometry
OTI2F9
Flow Cytometry
beta-lactamase-like protein 2, CGI-83, EC 3.-, lactamase, beta 2
Mouse
Affinity Purified
Primary
Human, Canine, Primate, Monkey
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.