Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
lactamase, beta 2 Antibody (OTI4F7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271903IR
Description
lactamase, beta 2 Monoclonal specifically detects lactamase, beta 2 in Human samples. It is validated for Western Blot.Specifications
lactamase, beta 2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LACTB2 | |
Full length recombinant protein of human lactamase, beta 2 (NP_057111) produced in E.coli. | |
0.1 mL | |
51110.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4F7 | |
Western Blot | |
beta-lactamase-like protein 2, CGI-83, EC 3.-, lactamase, beta 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction