Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactase Antibody (3C105.1), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250509MFV450
Description
Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| Lactase | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 3C105.1 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
| Normal human jejunal epithelial brush border-enriched membranes from a non-secretor and blood group O | |
| 0.1 mL | |
| Cancer, Cellular Signaling, metabolism | |
| 3938 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction