Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactase Antibody (3G5.1/2.G6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$173.10
Specifications
| Antigen | Lactase |
|---|---|
| Clone | 3G5.1/2.G6 |
| Concentration | 1 mg/mL |
| Dilution | Immunoprecipitation 1:10 - 1:500 |
| Applications | Immunoprecipitation |
Description
Lactase Monoclonal antibody specifically detects Lactase in Human, Rabbit samples. It is validated for ImmunoprecipitationSpecifications
| Lactase | |
| 1 mg/mL | |
| Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Signaling, metabolism | |
| PBS | |
| 3938 | |
| IgG1 | |
| Protein A purified |
| 3G5.1/2.G6 | |
| Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rabbit | |
| EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
| Normal jejunal epithelial brush border-enriched membranes from a non-secretor, blood group O | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title