Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactase Antibody (4E10 10.2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250513B
Description
Lactase Monoclonal specifically detects Lactase in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Lactase | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
LCT | |
Affinity purified human lactase | |
0.1 mL | |
Cancer, Cellular Signaling, metabolism | |
3938 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry | |
4E10 10.2 | |
Western Blot, Immunohistochemistry | |
EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction