Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Lactase Antibody (4E10 10.2), Janelia Fluor™ 646, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250513JF646
Description
Lactase Monoclonal specifically detects Lactase in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Lactase | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LCT | |
| Affinity purified human lactase | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified | 
| Western Blot, Immunohistochemistry | |
| 4E10 10.2 | |
| Western Blot, Immunohistochemistry | |
| EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
| Mouse | |
| Protein A purified | |
| Cancer, Cellular Signaling, metabolism | |
| 3938 | |
| Store at 4°C in the dark. | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction