Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactase-like Protein/LCTL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287714
Description
Lactase-like Protein/LCTL Polyclonal specifically detects Lactase-like Protein/LCTL in Human samples. It is validated for Western Blot.Specifications
Lactase-like Protein/LCTL | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
197021 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
FLJ33279, KLG, klotho gamma, Klotho/lactase-phlorizin hydrolase-related protein, KLPHKL lactase phlorizin hydrolase, lactase-like, lactase-like protein | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human Lactase-like Protein/LCTL | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction