Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactate Dehydrogenase Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306457MFV450
Description
Lactate Dehydrogenase Polyclonal antibody specifically detects Lactate Dehydrogenase in P. falciparum samples. It is validated for ELISASpecifications
Lactate Dehydrogenase | |
Polyclonal | |
ELISA | |
EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 | |
Produced in rabbits immunized with purified, recombinant Lactate Dehydrogenase (Accession#: XP_001349989.1; Met1-Ala316). | |
0.1 mL | |
HIF Target Genes, Hypoxia, Tumor Biomarkers | |
3945 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
P. falciparum | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction