Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactate Dehydrogenase B Antibody (14D7), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259441V
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase B Monoclonal antibody specifically detects Lactate Dehydrogenase B in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Lactate Dehydrogenase B | |
Monoclonal | |
DyLight 405 | |
EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 | |
Recombinant human Lactate Dehydrogenase B (1-334aa) purified from E. coli | |
0.1 mL | |
HIF Target Genes, Hypoxia, Mitochondrial Mediated Pathway | |
3945 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunofluorescence | |
14D7 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction