Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactate Dehydrogenase B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Lactate Dehydrogenase B |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 ug/ml, Flow Cytometry 2-5 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:400, Knockout Validated |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP253421SS
![]() |
Novus Biologicals
NBP253421SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP253421
![]() |
Novus Biologicals
NBP253421 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Lactate Dehydrogenase B Polyclonal specifically detects Lactate Dehydrogenase B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Lactate Dehydrogenase B | |
Western Blot 1 ug/ml, Flow Cytometry 2-5 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:400, Knockout Validated | |
Polyclonal | |
Rabbit | |
HIF Target Genes, Hypoxia, Mitochondrial Mediated Pathway | |
PBS with 0.02% Sodium Azide | |
3945 | |
Partial recombinant human Lactate Dehydrogenase B protein (amino acids 1-93) [P07195] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 | |
LDHB | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title