Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactate Dehydrogenase Polyclonal Antibody, Biotin, Rockland™
SDP

Catalog No. 501990485
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0485 25 μL
50-199-0484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0485 Supplier Rockland Immunochemicals Supplier No. 2001673S
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-lactate dehydrogenase antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Goat Serum as well as purified and partially purified Lactate Dehydrogenase [Rabbit Muscle]. Cross reactivity against Lactate Dehydrogenase from other sources may occur but have not been specifically determined.

Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84-89% sequence similarities and 69-75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure. The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers. Mutations in the LDHA gene can lead to Glycogen storage disease 11 (GSD11).

Specifications

Antigen Lactate Dehydrogenase
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Gene LDHA
Gene Accession No. P13491
Gene Alias Cell proliferation-inducing gene 19 protein; epididymis secretory sperm binding protein Li 133P; GSD11; HEL-S-133P; l7R2; Lactate Dehydrogenase; lactate dehydrogenase 1, A chain; lactate dehydrogenase A; lactate dehydrogenase A4; lactate dehydrogenase A-like; lactate dehydrogenase M; lactate dehydrogenase-A; lactate dehydrogenase-M; Lactic Dehydrogenase; LDH; LDH muscle subunit; Ldh1; Ldh-1; ldha; LDH-A; ldha protein; ldha1; LDH-A4 muscle-specific isozyme; glycolytic enzyme; LDHM; LDH-M; L-lactate dehydrogenase; L-lactate dehydrogenase A chain; M-LDH; OTTHUMP00000165206; PIG19; proliferation-inducing gene 19; Renal carcinoma antigen NY-REN-59
Gene Symbols LDHA
Host Species Goat
Immunogen Lactate Dehydrogenase [full length protein isolated from rabbit muscle]
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009107
Target Species Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.