Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Lactate-Glo Assay, 5ml

Catalog No. PRJ5021 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRJ5021 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRJ5021 Supplier Promega Supplier No. J5021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The Lactate-Glo Assay is a bioluminescent assay for rapid, selective and sensitive detection of L-lactate in biological samples.

  • Quick, Sensitive Measurement of Lactate from a Variety of Biological Samples
  • Perform in high−throughput workflows
  • Multiplex with other metabolite or viability assays
  • Linear range up to 200μM

Specifications

Content And Storage LESS THAN -65°C
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.