Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LACTB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557626 Supplier Invitrogen™ Supplier No. PA557626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSNENLLPGY LKPETMVMMW TPVPNTEMSW DKEGKYAMAW GVVEKKQTYG SCRKQRHYAS HTGGAVGASS VLLVLPEELD TETINNKVP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.

This gene encodes a protein from the large 39S subunit of the mitochondrial ribosome (mitoribosome). The encoded protein has some sequence similarity to prokaryotic beta-lactamases but most of the residues that are responsible for the beta-lactamase activity are not conserved between the two proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LACTB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LACTB
Gene Accession No. P83111
Gene Alias G24; Lact1; LACT-1; lactamase beta; lactamase, beta; Lactb; mitochondrial 39S ribosomal protein L56; mitochondrial ribosomal protein L56; MRPL56; serine beta lactamase-like protein; serine beta-lactamase-like protein LACTB, mitochondrial; UNQ843/PRO1781
Gene Symbols LACTB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LACTB. Recombinant protein control fragment (Product #RP-95825).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.