Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LACTB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117089 Supplier Invitrogen™ Supplier No. PA5117089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SKOV3, Mouse testis Immunogen sequence: DEVGAPGIVV GVSVDGKEVW SEGLGYADVE NRVPCKPETV MRIASISKSL TMVALAKLWE AGKLDLDIPV QHYVPEFPEK EYEGEKVSVT TRLLISHLSG IRHYEKDIKK VKEEKAYKAL KMMKENVAFE QEKEGKSNEK NDFTKFKTEQ ENEAKCRNSK PGKKKNDFEQ G.

This gene encodes a protein from the large 39S subunit of the mitochondrial ribosome (mitoribosome). The encoded protein has some sequence similarity to prokaryotic beta-lactamases but most of the residues that are responsible for the beta-lactamase activity are not conserved between the two proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LACTB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LACTB
Gene Accession No. P83111, Q9EP89
Gene Alias G24; Lact1; LACT-1; lactamase beta; lactamase, beta; Lactb; mitochondrial 39S ribosomal protein L56; mitochondrial ribosomal protein L56; MRPL56; serine beta lactamase-like protein; serine beta-lactamase-like protein LACTB, mitochondrial; UNQ843/PRO1781
Gene Symbols LACTB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-290 of human LACTB (NP_1162462).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114294, 80907
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.