Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LACTB2 Mouse anti-Human, Clone: OTI1A4, lyophilized, TrueMAB™
SDP

Catalog No. 501661462
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-1462 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-1462 Supplier OriGene Supplier No. CF502581
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Endoribonuclease; cleaves preferentially 3′ to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3′ -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.

Specifications

Antigen LACTB2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1A4
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene LACTB2
Gene Accession No. Q53H82
Gene Alias CGI-83
Gene Symbols LACTB2
Host Species Mouse
Immunogen Full length human recombit protein of human LACTB2 produced in E.coli
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51110
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.