Learn More
LACTB2 Mouse anti-Human, Clone: OTI4E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502583

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Endoribonuclease; cleaves preferentially 3′ to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3′ -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.Specifications
LACTB2 | |
Monoclonal | |
Unconjugated | |
LACTB2 | |
CGI-83 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51110 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI4E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q53H82 | |
LACTB2 | |
Full length human recombit protein of human LACTB2 produced in E.coli | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.