Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactic Acid, 85%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

50-21-5
C3H6O3
90.078
JVTAAEKCZFNVCJ-UHFFFAOYSA-N
612
CHEBI:78320
2-hydroxypropanoic acid
CC(C(=O)O)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 90.08
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.20kg
Quantity 500mL
Flash Point 112.00°C
Chemical Name or Material Lactic Acid
Assay 85%
Grade Baker Analyzed™ ACS Reagent