Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactic Acid, ACS, 85-90%, Spectrum™ Chemical
SDP

Catalog No. 18611179 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 mL
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Drum
Glass Bottle
Poly Bottle
Poly Drum

Chemical Identifiers

CAS 50-21-5

Specifications

CAS Max % 1
Percent Purity 85 to 90%
Grade ACS
Quantity 2.5 L
Packaging Glass Bottle
Residue after Ignition 0.0002

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.