Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Petrifilm™ Lactic Acid Bacteria Count Plate
SCI_ED

Catalog No. S30699 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
50 Pack
500 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30699 500 Pack
S30733 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30699 Supplier 3M™ Petrifilm™ Supplier No. 6462
Add to Cart
Add to Cart

Quickly and reliably detect lactic acid bacteria with sample-ready count plates. No prep, gas packs, or CO2 chambers required.

  • Product Type: Count Plate
  • Test Type: Environmental and Food Sample Test
  • Application: Detects total lactic acid bacteria populations in food and environmental samples
  • Incubation Time: Results in 48 hours
  • Format: Sample-ready plates – no media preparation needed
  • Anaerobic Testing: Built-in oxygen-scavenging and barrier films create self-contained anaerobic environment
  • No Additional Equipment Needed: Eliminates the need for gas packs, chambers, or CO2 incubators
  • PTM #041701 for selected foods
  • NF VALIDATION 3M 01/19-11/17 by AFNOR Certification (excluding yogurts)
  • Versatility: Compatible with a broad range of sample types and proven against MRS/APT agar methods
  • Pack Size: 500 plates

Specifications

Detects Total lactic acid bacteria populations
Product Type Count Plate
Test Type Environmental Test
For Use With (Application) Food and environmental samples
Quantity 500 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.