Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactic acid, lithium salt, 99%, extra pure

Catalog No. AC413335000
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413335000 500 g Glass bottle
AC413330050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413335000 Supplier Thermo Scientific Chemicals Supplier No. 413335000

CAS: 867-55-0 | C3H5LiO3 | 96.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-55-0
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
MDL Number MFCD00065399
InChI Key GKQWYZBANWAFMQ-UHFFFAOYSA-M
Synonym lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate
PubChem CID 23673452
IUPAC Name lithium;2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Absorbance 10% aq. soln., 10% aq. soln. at 0nm, 0.07 max. at 260nm
Color White
pH 6 to 9 (1% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Linear Formula CH3CH(OH)CO2Li
Quantity 500 g
Beilstein 03, IV, 633
Formula Weight 96.01
Percent Purity ≥98.5%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material Lactic acid, lithium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.