Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Antibody (002), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290704G
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Mouse samples. It is validated for ELISA.Specifications
Lactoferrin | |
Monoclonal | |
DyLight 488 | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Lactoferrin (AAH06904.1; Met1-Gln707). | |
0.1 mL | |
Amino Acids Drugs and other small molecules | |
4057 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
2 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction