Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Antibody (103), DyLight 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289795UV
Description
Lactoferrin Monoclonal antibody specifically detects Lactoferrin in Human samples. It is validated for ELISA, ELISASpecifications
Lactoferrin | |
Monoclonal | |
DyLight 350 | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Lactoferrin (P02788; Met1-Lys710). | |
0.1 mL | |
Amino Acids Drugs and other small molecules | |
4057 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
103 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction