Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Antibody (1C6cc), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010109H
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA.Specifications
Lactoferrin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LTF | |
Full length native protein (purified) (Human). | |
0.2 mL | |
Amino Acids Drugs and other small molecules | |
4057 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1C6cc | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human lactoferrin. No cross-reactivity with human transferrin. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction