Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Antibody (1C6cc), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010109
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA, CyTOF-ready.Specifications
Lactoferrin | |
Monoclonal | |
Unconjugated | |
P02788 | |
LTF | |
Full length native protein (purified) (Human). | |
Protein A purified | |
RUO | |
Primary | |
Human lactoferrin. No cross-reactivity with human transferrin. | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA, CyTOF | |
1C6cc | |
Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA 1:100-1:2000, CyTOF-ready | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
Mouse | |
78 kDa | |
0.2 mg | |
Amino Acids Drugs and other small molecules | |
4057 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction