Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Antibody (a-bC-lobe), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311196
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Bovine samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunoassay.Specifications
Lactoferrin | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Immunohistochemistry-Paraffin, Immunoassay | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
Monoclonal antibody a-bC-lobe, anti bovine Lactoferrin (Lf) is highly specific for bovine Lactoferrin. | |
50 μg | |
Amino Acids Drugs and other small molecules | |
4057 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin), Immunoassay | |
a-bC-lobe | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction