Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactoferrin Antibody (LF5-1D2), PE, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP267260PE

Catalog No. NBP267260PE


Only null left
Add to Cart

Description

Description

Lactoferrin Monoclonal specifically detects Lactoferrin in Human samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications

Specifications

Lactoferrin
Monoclonal
1mg/mL
Flow Cytometry, Flow (Intracellular)
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Mouse
Protein A purified
Amino Acids Drugs and other small molecules
4057
Store at 4°C in the dark.
IgG1
Flow Cytometry
LF5-1D2
PE
PBS with 0.05% Sodium Azide
LTF
Human lactoferrin
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.