Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Antibody (SR2256), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Lactoferrin |
---|---|
Clone | SR2256 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
Lactoferrin Monoclonal antibody specifically detects Lactoferrin in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
Lactoferrin | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
A synthesized peptide derived from human Lactoferrin (Uniprot #: P02788) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2256 | |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
4057 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title