Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lactoferrin Monoclonal Antibody (5F12.1.2)
GREENER_CHOICE

Catalog No. 01676385
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-385 Supplier Invitrogen™ Supplier No. MA140262
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-40262 detects Lactoferricin from bovine samples. MA1-40262 has been successfully used in ELISA and Western blot applications. The MA1-40262 immunogen is QWR region of Bovine Lactoferricin.

Lactoferrin (LF) is an iron binding glycoprotein. LF consists of a single polypeptide chain (approximately 80 kDa) folded into two structurally homologous lobes, each of which can reversibly bind one ferric ion (Fe3+). LF is found in external fluids, including milk and mucosal secretions, and is a prominent component of secondary granuals of neutrophils. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. LTF acts as a major first line defense against microbial infections, partly due to its ability to efficiently bind to and hence remove Fe3+ from the environment, but studies have also shown that LTF possesses bactericidal properties and proteolytic activity, capable of cleaving arginine-rich sequences within microbial proteins. Three isoforms of LTF have been identified: LTF-alpha, LTF-beta and LTF-gamma and receptors for LTF are expressed in the gastrointestinal tract, by macrophages, lymphocytes, neutrophils and platelets and also by some bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lactoferrin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5F12.1.2
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene LTF
Gene Accession No. P24627
Gene Alias Bovine Lactoferrin; CKRX; Csp82; epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HEL110; HLF2; Kaliocin 1; Kaliocin1; kaliocin-1; lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; LfcinH; Lfcin-H; LTF; minisatellite 10r detected by probe MMS10; MMS10R; Ms10r; N lobe; neutrophil lactoferrin; talalactoferrin
Gene Symbols LTF
Host Species Mouse
Immunogen QWR region of Bovine Lactoferricin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280846
Target Species Bovine
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.