Learn More
Invitrogen™ Lactoferrin Monoclonal Antibody (KT12)

Description
This product was used in both Indirect and Sandwich ELISA.
Specifications
Specifications
| Antigen | Lactoferrin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | KT12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | LTF |
| Gene Accession No. | P02788 |
| Gene Alias | Bovine Lactoferrin; CKRX; Csp82; epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HEL110; HLF2; Kaliocin 1; Kaliocin1; kaliocin-1; lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; LfcinH; Lfcin-H; LTF; minisatellite 10r detected by probe MMS10; MMS10R; Ms10r; N lobe; neutrophil lactoferrin; talalactoferrin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.