Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lactoferrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529744 Supplier Invitrogen™ Supplier No. PA529744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: human milk. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Lactoferrin (LF) is an iron binding glycoprotein. LF consists of a single polypeptide chain (approximately 80 kDa) folded into two structurally homologous lobes, each of which can reversibly bind one ferric ion (Fe3+). LF is found in external fluids, including milk and mucosal secretions, and is a prominent component of secondary granuals of neutrophils. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. LTF acts as a major first line defense against microbial infections, partly due to its ability to efficiently bind to and hence remove Fe3+ from the environment, but studies have also shown that LTF possesses bactericidal properties and proteolytic activity, capable of cleaving arginine-rich sequences within microbial proteins. Three isoforms of LTF have been identified: LTF-alpha, LTF-beta and LTF-gamma and receptors for LTF are expressed in the gastrointestinal tract, by macrophages, lymphocytes, neutrophils and platelets and also by some bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lactoferrin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene LTF
Gene Accession No. P02788
Gene Alias Bovine Lactoferrin; CKRX; Csp82; epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HEL110; HLF2; Kaliocin 1; Kaliocin1; kaliocin-1; lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; LfcinH; Lfcin-H; LTF; minisatellite 10r detected by probe MMS10; MMS10R; Ms10r; N lobe; neutrophil lactoferrin; talalactoferrin
Gene Symbols LTF
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Lactoferrin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4057
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.