Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoferrin Rabbit anti-Mouse, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290704100UL
Description
Lactoferrin Monoclonal specifically detects Lactoferrin in Mouse samples. It is validated for ELISA.Specifications
| Lactoferrin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 2 | |
| ELISA 1:5000-1:10000 | |
| EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin | |
| Recombinant Mouse Lactoferrin protein | |
| 100 μg | |
| Amino Acids Drugs and other small molecules | |
| 4057 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction