Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoperoxidase Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11061574AF700
Description
Lactoperoxidase Polyclonal specifically detects Lactoperoxidase in Human, Mouse, Bovine, Goat, Sheep samples. It is validated for Western Blot.Specifications
Lactoperoxidase | |
Polyclonal | |
Western Blot | |
P22079 | |
LPO | |
Synthetic peptide made to a C-terminal portion of the human Lactoperoxidase protein (within residues 650-712). [Swiss-Prot# P22079] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Bovine, Goat, Sheep | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 1.11.1, lactoperoxidase, MGC129990, MGC129991, Salivary peroxidase, SAPX, SPOEC 1.11.1.7 | |
Rabbit | |
100 kDa | |
0.1 mL | |
Cancer, Immunology, Virology Bacteria and Parasites | |
4025 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction