Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoperoxidase Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11061572AF750
Description
Lactoperoxidase Polyclonal specifically detects Lactoperoxidase in Human, Bovine, Primate samples. It is validated for Western Blot.Specifications
| Lactoperoxidase | |
| Polyclonal | |
| Western Blot | |
| EC 1.11.1, lactoperoxidase, MGC129990, MGC129991, Salivary peroxidase, SAPX, SPOEC 1.11.1.7 | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Immunology, Virology Bacteria and Parasites | |
| 4025.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LPO | |
| Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400). [Swiss-ProtNo. P22079] | |
| 0.1 mL | |
| Primary | |
| Human, Bovine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction