Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactoperoxidase Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11061572B
Description
Lactoperoxidase Polyclonal specifically detects Lactoperoxidase in Human, Bovine, Primate samples. It is validated for Western Blot.Specifications
Lactoperoxidase | |
Polyclonal | |
Western Blot | |
P22079 | |
LPO | |
Synthetic peptide made to an internal portion of the human Lactoperoxidase protein (within residues 300-400). [Swiss-Prot# P22079] | |
Affinity Purified | |
RUO | |
Primary | |
Human, monkey and bovine. Immunogen has 81% homology to sheep and 73% homology to mouse and rat. | |
Store at 4C in the dark. |
Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
EC 1.11.1, lactoperoxidase, MGC129990, MGC129991, Salivary peroxidase, SAPX, SPOEC 1.11.1.7 | |
Rabbit | |
100 kDa | |
0.1 mL | |
Cancer, Immunology, Virology Bacteria and Parasites | |
4025 | |
Human, Bovine, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction