Learn More
Lactoperoxidase Polyclonal Antibody, Rockland™

Sheep Polyclonal Antibody
$178.35 - $649.44
Specifications
Antigen | Lactoperoxidase |
---|---|
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1249
![]() ![]() |
Rockland Immunochemicals
2006011460100 |
100 μg |
Each for $649.44
|
|
|||||
50-199-1250
![]() ![]() |
Rockland Immunochemicals
200601146S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-LACTOPEROXIDASE is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Sheep Serum as well as purified and partially purified Lactoperoxidase [Bovine Milk]. Cross reactivity against Lactoperoxidase from other tissues and species may occur but have not been specifically determined.
This gene encodes an oxidoreductase secreted from salivary, mammary, and other mucosal glands that functions as a natural antibacterial agent. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Lactoperoxidase | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
5830499B15Rik; airway lactoperoxidase; EC 1.11.17 antibody; Lactoperoxidase; lactoperoxydase; LPO; LPO antibody; MGC129990; MGC129991; Salivary peroxidase; SAPX; SAPX antibody; SPO; SPO antibody | |
LPO | |
Primary | |
Antibody |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Sheep | |
Bovine | |
P80025 | |
280844 | |
Lactoperoxidase [Bovine Milk] | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
LPO |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.