Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactophenol Cotton Blue, MilliporeSigma™
Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 mL
Description
Mounting fluid for demonstrating microscopic morphology of fungi
Specifications
Specifications
| Chemical Name or Material | Lactophenol Cotton Blue |
| Name Note | Fungi Stain |
| Packaging | Poly Bottle |
| Quantity | 100 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction