Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lactose, Monohydrate, Spray-Dried Powder, NF, Spectrum™ Chemical
SDP

Catalog No. 18603699 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

LA105, 64044-51-5, C12H22O11·H2O

Spectrum™ Chemical Lactose, Monohydrate, Spray-Dried Powder, NF is used to help form tablets because it has excellent compressibility properties and as a diluent powder for dry-powder inhalations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
InChI Key WSVLPVUVIUVCRA-RJMJUYIDSA-N
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
SMILES O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Poly Pail
Quantity 2.5 kg
Specific Rotation 110.3
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.