Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lactyl-Lysine Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $504.00
Specifications
Antigen | Lactyl-Lysine |
---|---|
Dilution | Western Blot 1:500 - 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Lactyl-Lysine Polyclonal antibody specifically detects Lactyl-Lysine in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Lactyl-Lysine | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Kla, Lactylated Lysine, Lactyllysine | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
PBS, 50% glycerol, pH7.3 | |
A synthetic peptide corresponding to a sequence containing lactated K | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title