Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120555
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120555 Supplier Invitrogen™ Supplier No. PA5120555
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa. Immunogen sequence: TTDDEAPRLS QNGDRQASAS ERLPSVEEAE VPKPLPPASK DEDEDIQSIL RTRQERRQRR QVVEAAQAPI QERLEAEEGR NSLSPVQATQ KPLVSKKELE IPPRRRLSRE QRGPWALEEE SLVGREPEER KKGVPEKSPV LEKSSMPKKT APEKSLVSDK T.

Anchoring filament protein which is a component of the basement membrane zone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LAD1
Gene Accession No. O00515, P57016
Gene Alias LAD; Lad1; Lad-1; LADA; ladinin; ladinin 1; ladinin-1; linear IgA bullous dermatosis antigen; Linear IgA disease antigen; Linear IgA disease antigen homolog
Gene Symbols LAD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-200 of human LAD1 (NP_005549.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16763, 313325, 3898
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.