Learn More
Description
Immunohistochemical staining of 16136-1-AP shows strong extracellular matrixpositivity in human testis. The predicted MW of this protein is 57 kDa. IP results of 16136-1-AP detected a 62 kDa band.
Specifications
Specifications
| Antigen | LAD1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LAD1 |
| Gene Accession No. | O00515, P57016 |
| Gene Alias | LAD, Lad 1, LAD1, LadA, ladinin 1, Linear IgA disease antigen |
| Gene Symbols | LAD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
