Learn More
PIP™ Lady Eva Reader Bifocal
Catalog No. 19148258 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Magnification Power:
+1.00
+1.25
+1.50
+1.75
+2.00
+2.25
+2.50
+2.75
+3.00
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-148-258 +1.75
19-148-255 +1.00
19-148-257 +1.50
19-148-261 +2.50
19-148-256 +1.25
19-148-259 +2.00
19-148-260 +2.25
19-148-262 +2.75
19-148-263 +3.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-148-258 Supplier PIP™ Supplier No. 250120175
This item is not returnable. View return policy
PIP™ Lady Eva Reader Bifocal features a construction that better fits female facial structure
  • Designed for the worker who needs to see up close
  • Offered in increments of +0.25 diopters to give users the exact reading power of their personal glasses
  • Lightweight, rimless design provides all day user comfort
  • Rubber co-molded temples provide a secure fit
  • Molded nose bridge design to fit the vast majority of users
  • 10 Base curve lens
  • Meets ANSI Z87.1+
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
Frame Color Clear, Pink
Magnification Power +1.75
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Rimless
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Warning - Prop 65 - Reproductive Harm