Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 Antibody (1009614), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB23194H
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| LAG-3 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity | |
| 3902 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 1009614 | |
| Flow Cytometry, CyTOF-ready | |
| CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
| Mouse myeloma cell line NS0-derived recombinant human LAG-3, Met1-Leu450, Accession # P18627 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction