Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 Antibody (11E3), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197662APCCY7
Description
LAG-3 Monoclonal specifically detects LAG-3 in Human, Monkey samples. It is validated for Flow Cytometry.Specifications
LAG-3 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
LAG3 | |
Recombinant human LAG-3 (lymphocyte activation gene-3). | |
0.1 mL | |
Primary | |
Recognizes the first N-terminal D1 domain of human and monkey LAG-3. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
11000 | |
Flow Cytometry | |
CD223, CD223 antigen, FDC, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Immunology, Innate Immunity | |
3902 | |
Human, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction