Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG-3 Antibody (17B4), Novus Biologicals™
SDP

Catalog No. NB258731 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258731 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB258731 Supplier Novus Biologicals Supplier No. NBP280059
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Inhibition Activity

Specifications

Antigen LAG-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Inhibition Assays
Classification Monoclonal
Clone 17B4
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 5 ug, Flow Cytometry, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Frozen 1:150, Inhibition Activity
Formulation PBS
Gene Alias CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
Host Species Mouse
Immunogen Synthetic peptide corresponding to 30 aa in the N-terminus of human LAG-3.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3902
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.