Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    LAG-3 Antibody (1C6) - VHH - Azide and BSA Free, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Llama Monoclonal Antibody
Supplier: Novus Biologicals NBP312839
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| LAG-3 | |
| Monoclonal | |
| 1 mg/ml | |
| PBS | |
| Llama | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1C6 | |
| Unconjugated | |
| CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
| LAG-3 antibody was raised against a recombinant protein corresponding to amino acids 23-434 of human LAG-3. | |
| 0.1 mg | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity | |
| 3902 | |
| Store at -20°C. Avoid freeze-thaw cycles. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction