Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 Antibody (631501) [Janelia Fluor™ 646], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB33281J
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
LAG-3 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
631501 | |
Flow Cytometry, CyTOF-ready | |
CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
NS0 mouse myeloma cell line transfected with mouse LAG-3, Ser23-Leu442, Accession # Q61790 | |
0.1 mL | |
Adaptive Immunity, Cancer, Immunology, Innate Immunity | |
3902 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction