Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 Antibody (874525), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB23191JF549
Description
LAG-3 Monoclonal specifically detects LAG-3 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
LAG-3 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LAG3 | |
Mouse myeloma cell line NS0-derived partial recombinant human LAG-3 protein (amino acids 23-450) | |
0.1 mL | |
Adaptive Immunity, Immunology, Innate Immunity | |
Human | |
IgG2a |
Western Blot, Flow Cytometry | |
874525 | |
Western Blot, Flow Cytometry | |
CD223, CD223 antigen, FDC, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction